The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308006.1 | NP_001294935.1 | 2 - 3 | 561 | 88 |
NM_006033.3 | NP_006024.1 | 2 - 3 | 561 | 88 | |
XM_005258390.1 | XP_005258447.1 | 2 - 3 | 1378 | 88 | |
XM_011526265.2 | XP_011524567.1 | 2 - 3 | 591 | 88 | |
XM_011526267.1 | XP_011524569.1 | 2 - 3 | 948 | 88 | |
XM_017026095.1 | XP_016881584.1 | 2 - 3 | 948 | 88 | |
GenBank mRNA | AF118767.1 | - | 2 - 3 | 535 | 88 |
AK125344.1 | - | 2 - 3 | 1378 | 88 | |
AK291799.1 | - | 2 - 3 | 535 | 88 | |
AK300333.1 | - | 2 - 3 | 532 | 88 | |
AY358928.1 | - | 2 - 3 | 505 | 88 | |
BC060825.1 | - | 2 - 3 | 532 | 88 |