Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD1
Entrez Gene ID
953
Gene Name
ectonucleoside triphosphate diphosphohydrolase 1
Gene Aliases
ATPDase, CD39, NTPDase-1, SPG64
Chromosome Location
Chr.10: 95710901 - 95877266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098175.1 NP_001091645.1 6 - 7 1014 68
NM_001164178.1 NP_001157650.1 6 - 7 914 68
NM_001164181.1 NP_001157653.1 5 - 6 786 68
NM_001164182.1 NP_001157654.1 5 - 6 988 68
NM_001164183.1 NP_001157655.1 4 - 5 870 68
NM_001312654.1 NP_001299583.1 6 - 7 960 68
NM_001320916.1 NP_001307845.1 6 - 7 914 68
NM_001776.5 NP_001767.3 6 - 7 1139 68
XM_011540370.2 XP_011538672.2 5 - 6 874 68
XM_011540371.2 XP_011538673.1 7 - 8 1150 68
XM_011540372.2 XP_011538674.2 5 - 6 921 68
XM_011540373.2 XP_011538675.2 5 - 6 917 68
XM_011540374.2 XP_011538676.2 5 - 6 889 68
XM_011540376.2 XP_011538678.2 5 - 6 940 68
XM_011540377.2 XP_011538679.1 6 - 7 1021 68
XM_017016958.1 XP_016872447.1 6 - 7 1281 68
XM_017016959.1 XP_016872448.1 4 - 5 604 68
XM_017016960.1 XP_016872449.1 5 - 6 760 68
XM_017016961.1 XP_016872450.1 5 - 6 745 68
GenBank mRNA AJ133133.1 - 6 - 7 1019 68
AK298648.1 - 6 - 7 885 68
AK301459.1 - 6 - 7 903 68
AK304018.1 - 6 - 7 900 68
AK304836.1 - 5 - 6 742 68
AK316009.1 - 5 - 6 691 68
BC047664.1 - 4 - 5 607 68
S73813.1 - 6 - 7 882 68
U87967.1 - 6 - 7 845 68

View Full Product Details