The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001686.3 | NP_001677.2 | 3 - 4 | 587 | 96 |
GenBank mRNA | AK291085.1 | - | 3 - 4 | 503 | 96 |
BC016512.1 | - | 3 - 4 | 492 | 96 | |
DQ403107.1 | - | 2 - 3 | 290 | 96 | |
FJ224288.1 | - | 3 - 4 | 587 | 96 |