Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5B
Entrez Gene ID
506
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
ATPMB, ATPSB, HEL-S-271
Chromosome Location
Chr.12: 56638175 - 56646068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001686.3 NP_001677.2 4 - 5 709 81
GenBank mRNA AK291085.1 - 4 - 5 625 81
BC016512.1 - 4 - 5 614 81
DQ403107.1 - 3 - 4 412 81
FJ224288.1 - 4 - 5 709 81

View Full Product Details