Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNOT2
Entrez Gene ID
4848
Gene Name
CCR4-NOT transcription complex subunit 2
Gene Aliases
CDC36, HSPC131, NOT2, NOT2H
Chromosome Location
Chr.12: 70242993 - 70354993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199302.1 NP_001186231.1 11 - 12 1469 116
NM_001199303.1 NP_001186232.1 10 - 11 1722 116
NM_014515.5 NP_055330.1 10 - 11 1600 116
NR_037615.1 11 - 12 1946 116
XM_006719429.3 XP_006719492.1 11 - 12 1825 116
XM_006719430.3 XP_006719493.1 11 - 12 1659 116
XM_006719432.3 XP_006719495.1 9 - 10 1418 116
XM_006719433.3 XP_006719496.1 7 - 8 1190 116
XM_011538399.1 XP_011536701.1 10 - 11 1555 116
XM_011538400.2 XP_011536702.1 11 - 12 1788 116
XM_017019348.1 XP_016874837.1 12 - 13 1846 116
XM_017019349.1 XP_016874838.1 12 - 13 2012 116
XM_017019350.1 XP_016874839.1 12 - 13 1780 116
XM_017019351.1 XP_016874840.1 12 - 13 1946 116
XM_017019352.1 XP_016874841.1 13 - 14 1967 116
XM_017019353.1 XP_016874842.1 8 - 9 1248 116
XM_017019354.1 XP_016874843.1 8 - 9 1414 116
GenBank mRNA AF044215.1 - 10 - 11 1157 116
AF113226.1 - 9 - 10 1127 116
AF161480.1 - 10 - 11 1138 116
AF180473.1 - 10 - 11 1133 116
AK000662.1 - 10 - 11 1013 116
AK095789.1 - 10 - 11 1685 116
AK315715.1 - 11 - 12 1470 116
AL137674.1 - 2 - 3 149 116
BC002597.2 - 10 - 11 1144 116
BC011826.2 - 10 - 11 1152 116
BX641116.1 - 1 - 2 3524 116

View Full Product Details