The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145166.1 | NP_001138638.1 | 13 - 14 | 1518 | 96 |
NM_001145167.1 | NP_001138639.1 | 14 - 15 | 1568 | 96 | |
NM_005788.3 | NP_005779.1 | 15 - 16 | 1704 | 96 | |
XM_011519836.2 | XP_011518138.1 | 12 - 13 | 1681 | 96 | |
XM_017017081.1 | XP_016872570.1 | 14 - 15 | 1440 | 96 | |
GenBank mRNA | AF059531.1 | - | 14 - 15 | 1431 | 96 |
AK125039.1 | - | 7 - 8 | 3181 | 96 | |
AK129984.1 | - | 14 - 15 | 1622 | 96 | |
AK298185.1 | - | 14 - 15 | 1382 | 96 | |
AK300591.1 | - | 13 - 14 | 1332 | 96 | |
AK307776.1 | - | 11 - 12 | 1163 | 96 | |
BC019339.1 | - | 14 - 15 | 1540 | 96 | |
BC037544.1 | - | 15 - 16 | 1492 | 96 | |
BC064831.1 | - | 15 - 16 | 1679 | 96 |