The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145166.1 | NP_001138638.1 | 7 - 8 | 915 | 159 |
NM_001145167.1 | NP_001138639.1 | 8 - 9 | 965 | 159 | |
NM_005788.3 | NP_005779.1 | 9 - 10 | 1101 | 159 | |
XM_011519836.2 | XP_011518138.1 | 6 - 7 | 1078 | 159 | |
XM_017017081.1 | XP_016872570.1 | 8 - 9 | 837 | 159 | |
GenBank mRNA | AF059531.1 | - | 8 - 9 | 828 | 159 |
AK125039.1 | - | 1 - 2 | 2578 | 159 | |
AK129984.1 | - | 8 - 9 | 1019 | 159 | |
AK298185.1 | - | 8 - 9 | 779 | 159 | |
AK300591.1 | - | 7 - 8 | 729 | 159 | |
AK301449.1 | - | 9 - 10 | 933 | 159 | |
AK307776.1 | - | 5 - 6 | 560 | 159 | |
BC019339.1 | - | 8 - 9 | 937 | 159 | |
BC037544.1 | - | 9 - 10 | 889 | 159 | |
BC064831.1 | - | 9 - 10 | 1076 | 159 |