The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297709.1 | NP_001284638.1 | 1 - 2 | 255 | 102 |
NM_001297711.1 | NP_001284640.1 | 1 - 2 | 255 | 102 | |
NM_001297712.1 | NP_001284641.1 | 1 - 2 | 255 | 102 | |
NM_003480.3 | NP_003471.1 | 1 - 2 | 255 | 102 | |
NR_123733.1 | 1 - 2 | 255 | 102 | ||
NR_123734.1 | 1 - 2 | 255 | 102 | ||
GenBank mRNA | AK124368.1 | - | 1 - 2 | 251 | 102 |
AK290585.1 | - | 1 - 2 | 46 | 102 | |
AK299475.1 | - | 1 - 2 | 83 | 102 | |
AK315807.1 | - | 1 - 2 | 42 | 102 | |
AU280458.1 | - | 1 - 2 | 193 | 102 | |
BC005901.1 | - | 1 - 2 | 45 | 102 | |
BU929788.1 | - | 1 - 2 | 59 | 102 | |
DA544968.1 | - | 1 - 2 | 92 | 102 | |
DN996275.1 | - | 1 - 2 | 76 | 102 | |
U37283.1 | - | 1 - 2 | 177 | 102 |