The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297709.1 | NP_001284638.1 | 2 - 3 | 322 | 85 |
NM_001297711.1 | NP_001284640.1 | 2 - 3 | 322 | 85 | |
NM_001297712.1 | NP_001284641.1 | 2 - 3 | 322 | 85 | |
NM_003480.3 | NP_003471.1 | 2 - 3 | 322 | 85 | |
NR_123733.1 | 2 - 3 | 322 | 85 | ||
NR_123734.1 | 2 - 3 | 322 | 85 | ||
GenBank mRNA | AK124368.1 | - | 2 - 3 | 318 | 85 |
AK299475.1 | - | 2 - 3 | 150 | 85 | |
AK315807.1 | - | 2 - 3 | 109 | 85 | |
AU280458.1 | - | 2 - 3 | 260 | 85 | |
AY339060.1 | - | 1 - 2 | 58 | 85 | |
BC005901.1 | - | 2 - 3 | 112 | 85 | |
BU929788.1 | - | 2 - 3 | 126 | 85 | |
DA544968.1 | - | 2 - 3 | 159 | 85 | |
DB247952.1 | - | 2 - 3 | 322 | 85 | |
DN996275.1 | - | 2 - 3 | 143 | 85 | |
U37283.1 | - | 2 - 3 | 244 | 85 |