Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC10
Entrez Gene ID
54819
Gene Name
zinc finger CCHC-type containing 10
Gene Aliases
-
Chromosome Location
Chr.5: 132996985 - 133026621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300816.2 NP_001287745.1 4 - 5 384 91
NM_001308127.1 NP_001295056.1 4 - 5 414 91
NM_017665.3 NP_060135.1 3 - 4 318 91
NR_125337.2 3 - 4 322 91
GenBank mRNA AF086446.1 - 2 - 3 200 91
AK000101.1 - 3 - 4 262 91
BC005211.1 - 3 - 4 275 91
BC022443.1 - 4 - 5 325 91
BI826979.1 - 3 - 4 318 91
BM470321.1 - 3 - 4 269 91
DA905071.1 - 4 - 5 359 91

View Full Product Details