The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144002.1 | NP_001137474.1 | 18 - 19 | 2546 | 154 |
NM_019857.4 | NP_062831.3 | 18 - 19 | 2099 | 154 | |
NM_175859.2 | NP_787055.1 | 18 - 19 | 2185 | 154 | |
XM_005274562.3 | XP_005274619.1 | 18 - 19 | 1962 | 154 | |
XM_006724503.3 | XP_006724566.1 | 19 - 20 | 2137 | 154 | |
XM_011545545.2 | XP_011543847.1 | 18 - 19 | 1958 | 154 | |
GenBank mRNA | AF086422.1 | - | 1 - 2 | 154 | |
AF226667.1 | - | 18 - 19 | 2002 | 154 | |
AK023549.1 | - | 18 - 19 | 2057 | 154 | |
AK024070.1 | - | 18 - 19 | 2040 | 154 | |
AK025654.1 | - | 6 - 7 | 551 | 154 | |
AK125332.1 | - | 18 - 19 | 2546 | 154 | |
AK125348.1 | - | 18 - 19 | 2176 | 154 | |
AK309508.1 | - | 3 - 4 | 602 | 154 | |
BC006256.2 | - | 16 - 17 | 1541 | 154 | |
BC034986.1 | - | 18 - 19 | 2164 | 154 | |
BX647712.1 | - | 1 - 2 | 542 | 154 |