The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005505.4 | NP_005496.4 | 11 - 12 | 1653 | 86 |
GenBank mRNA | AF515445.1 | - | 12 - 13 | 1715 | 86 |
BC080647.1 | - | 11 - 12 | 1527 | 86 | |
BC093732.1 | - | 11 - 12 | 1463 | 86 | |
BC112037.1 | - | 11 - 12 | 1475 | 86 | |
Z22555.1 | - | 11 - 12 | 1469 | 86 |