Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBED5
Entrez Gene ID
58486
Gene Name
zinc finger BED-type containing 5
Gene Aliases
Buster1
Chromosome Location
Chr.11: 10852704 - 10858073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143667.1 NP_001137139.1 1 - 2 210 102
NM_021211.3 NP_067034.2 1 - 2 210 102
GenBank mRNA AF205600.1 - 1 - 2 179 102
AK315033.1 - 1 - 2 174 102
AL137577.1 - 1 - 2 934 102
BC017803.1 - 1 - 2 182 102
BC047754.1 - 1 - 2 178 102
DA059122.1 - 1 - 2 210 102
DA785416.1 - 1 - 2 210 102
DB100756.1 - 1 - 2 210 102

View Full Product Details