The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005288.3 | NP_005279.1 | 1 - 2 | 211 | 121 |
GenBank mRNA | AK290220.1 | - | 1 - 2 | 212 | 121 |
AK307756.1 | - | 1 - 2 | 211 | 121 |