Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UEVLD
Entrez Gene ID
55293
Gene Name
UEV and lactate/malate dehyrogenase domains
Gene Aliases
ATTP, UEV3
Chromosome Location
Chr.11: 18529609 - 18588837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040697.2 NP_001035787.1 4 - 5 447 151
NM_001261382.1 NP_001248311.1 3 - 4 381 151
NM_001261383.1 NP_001248312.1 3 - 4 381 151
NM_001261385.1 NP_001248314.1 4 - 5 404 151
NM_001297771.1 NP_001284700.1 4 - 5 447 151
NM_018314.4 NP_060784.3 4 - 5 447 151
XM_006718255.2 XP_006718318.1 3 - 4 313 151
GenBank mRNA AF503350.1 - 4 - 5 407 151
AK001930.1 - 4 - 5 404 151
AK314521.1 - 4 - 5 447 151
BC011011.1 - 4 - 5 384 151
BC064566.1 - 3 - 4 321 151
CB991602.1 - 4 - 5 462 151

View Full Product Details