The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198512.2 | NP_940914.1 | 3 - 4 | 368 | 102 |
GenBank mRNA | AK129500.1 | - | 3 - 4 | 365 | 102 |
BG696880.1 | - | 3 - 4 | 367 | 102 |