Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGAT2L6
Entrez Gene ID
347516
Gene Name
diacylglycerol O-acyltransferase 2 like 6
Gene Aliases
DC3
Chromosome Location
Chr.X: 70177483 - 70205703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198512.2 NP_940914.1 4 - 5 575 70
GenBank mRNA AK129500.1 - 4 - 5 572 70
BG696880.1 - 4 - 5 574 70

View Full Product Details