Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
Chromosome Location
Chr.1: 111227659 - 111243440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025197.1 NP_001020368.1 5 - 6 644 87
NM_001025199.1 NP_001020370.1 5 - 6 704 87
NM_004000.2 NP_003991.2 6 - 7 674 87
GenBank mRNA AK223316.1 - 5 - 6 584 87
AK298466.1 - 5 - 6 595 87
BC011460.1 - 6 - 7 612 87
BQ181849.1 - 4 - 5 87
BT006767.1 - 6 - 7 603 87
U49835.1 - 6 - 7 623 87
U58514.1 - 6 - 7 639 87
U58515.1 - 5 - 6 705 87

View Full Product Details