The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014443.2 | NP_001014443.1 | 11 - 12 | 1762 | 90 |
NM_001319848.1 | NP_001306777.1 | 10 - 11 | 1621 | 90 | |
NM_012475.4 | NP_036607.3 | 10 - 11 | 1621 | 90 | |
XM_017001040.1 | XP_016856529.1 | 10 - 11 | 1684 | 90 | |
GenBank mRNA | AF177758.1 | - | 10 - 11 | 1472 | 90 |
AF217979.1 | - | 10 - 11 | 1545 | 90 | |
AF233442.1 | - | 10 - 11 | 1559 | 90 | |
AK292319.1 | - | 11 - 12 | 1707 | 90 | |
AK310326.1 | - | 10 - 11 | 1654 | 90 | |
AL157417.1 | - | 6 - 7 | 1937 | 90 | |
BC003130.1 | - | 9 - 10 | 1121 | 90 | |
BC090946.1 | - | 11 - 12 | 1710 | 90 | |
BC136291.1 | - | 9 - 10 | 1409 | 90 |