The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194990.1 | NP_001181919.1 | 4 - 5 | 1677 | 60 |
NM_001194991.1 | NP_001181920.1 | 4 - 5 | 1262 | 60 | |
NM_001194992.1 | NP_001181921.1 | 4 - 5 | 1127 | 60 | |
NM_019606.5 | NP_062552.2 | 3 - 4 | 2410 | 60 | |
XM_011516410.2 | XP_011514712.1 | 4 - 5 | 1735 | 60 | |
GenBank mRNA | AK000264.1 | - | 3 - 4 | 1065 | 60 |
AK055964.1 | - | 4 - 5 | 1650 | 60 | |
AK291366.1 | - | 3 - 4 | 2205 | 60 | |
BC000556.2 | - | 3 - 4 | 2135 | 60 | |
BC016396.1 | - | 3 - 4 | 2410 | 60 | |
BC018935.2 | - | 3 - 4 | 2135 | 60 |