Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GALNT7
Entrez Gene ID
51809
Gene Name
polypeptide N-acetylgalactosaminyltransferase 7
Gene Aliases
GALNAC-T7, GalNAcT7
Chromosome Location
Chr.4: 173168753 - 173323967 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017423.2 NP_059119.2 4 - 5 963 68
XM_005263062.1 XP_005263119.1 4 - 5 963 68
XM_011532025.2 XP_011530327.1 4 - 5 1710 68
XM_017008292.1 XP_016863781.1 7 - 8 1883 68
XM_017008293.1 XP_016863782.1 7 - 8 3672 68
GenBank mRNA AK295509.1 - 2 - 3 270 68
BC035303.1 - 3 - 4 394 68
BC046129.1 - 4 - 5 941 68
BC047468.1 - 4 - 5 938 68

View Full Product Details