The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032813.3 | NP_116202.2 | 2 - 3 | 405 | 73 |
XM_011521121.2 | XP_011519423.1 | 2 - 3 | 383 | 73 | |
XM_011521122.2 | XP_011519424.1 | 2 - 3 | 299 | 73 | |
XM_011521125.1 | XP_011519427.1 | 2 - 3 | 292 | 73 | |
XM_017020798.1 | XP_016876287.1 | 2 - 3 | 383 | 73 | |
XM_017020800.1 | XP_016876289.1 | 2 - 3 | 292 | 73 | |
GenBank mRNA | AK056409.1 | - | 2 - 3 | 265 | 73 |