The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193522.1 | NP_001180451.1 | 15 - 16 | 2866 | 139 |
NM_001193523.1 | NP_001180452.1 | 15 - 16 | 2979 | 139 | |
NM_001193524.1 | NP_001180453.1 | 15 - 16 | 2892 | 139 | |
NM_024519.3 | NP_078795.2 | 15 - 16 | 2857 | 139 | |
XM_011523321.1 | XP_011521623.1 | 15 - 16 | 3671 | 139 | |
XM_011523322.1 | XP_011521624.1 | 15 - 16 | 2866 | 139 | |
XM_011523324.2 | XP_011521626.1 | 15 - 16 | 2926 | 139 | |
XM_011523325.1 | XP_011521627.1 | 15 - 16 | 2881 | 139 | |
XM_011523326.1 | XP_011521628.1 | 15 - 16 | 3080 | 139 | |
XM_017023663.1 | XP_016879152.1 | 15 - 16 | 2863 | 139 | |
XM_017023664.1 | XP_016879153.1 | 14 - 15 | 2752 | 139 | |
GenBank mRNA | AB067517.1 | - | 14 - 15 | 2712 | 139 |
AK023787.1 | - | 10 - 11 | 2349 | 139 | |
AK127792.1 | - | 15 - 16 | 2866 | 139 | |
AK293748.1 | - | 15 - 16 | 2889 | 139 | |
AK295677.1 | - | 15 - 16 | 2823 | 139 | |
AK302868.1 | - | 5 - 6 | 2013 | 139 | |
AL834312.1 | - | 5 - 6 | 1922 | 139 | |
BC001850.4 | - | 1 - 2 | 132 | 139 | |
BC054512.1 | - | 5 - 6 | 1899 | 139 | |
BC098587.1 | - | 9 - 10 | 2294 | 139 |