Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
Chromosome Location
Chr.16: 67528814 - 67546788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193522.1 NP_001180451.1 19 - 20 3525 56
NM_001193523.1 NP_001180452.1 19 - 20 3638 56
NM_001193524.1 NP_001180453.1 19 - 20 3551 56
NM_024519.3 NP_078795.2 19 - 20 3516 56
XM_011523321.1 XP_011521623.1 19 - 20 4330 56
XM_011523322.1 XP_011521624.1 19 - 20 3525 56
XM_011523324.2 XP_011521626.1 19 - 20 3585 56
XM_011523325.1 XP_011521627.1 19 - 20 3540 56
XM_011523326.1 XP_011521628.1 19 - 20 3739 56
XM_017023663.1 XP_016879152.1 19 - 20 3522 56
XM_017023664.1 XP_016879153.1 18 - 19 3411 56
GenBank mRNA AB067517.1 - 18 - 19 3371 56
AK023787.1 - 14 - 15 3008 56
AK127792.1 - 19 - 20 3525 56
AK293748.1 - 19 - 20 3548 56
AK295677.1 - 19 - 20 3482 56
AK302868.1 - 9 - 10 2672 56
AL834312.1 - 9 - 10 2581 56
BC001850.4 - 5 - 6 791 56
BC054512.1 - 9 - 10 2558 56
BC098587.1 - 13 - 14 2953 56

View Full Product Details