The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100398.1 | NP_001093868.1 | 13 - 14 | 1243 | 53 |
NM_015085.4 | NP_055900.4 | 14 - 15 | 1288 | 53 | |
XM_005256542.2 | XP_005256599.1 | 13 - 14 | 1680 | 53 | |
XM_006721477.3 | XP_006721540.1 | 14 - 15 | 1229 | 53 | |
XM_011523738.1 | XP_011522040.1 | 15 - 16 | 1341 | 53 | |
XM_011523739.1 | XP_011522041.1 | 14 - 15 | 1296 | 53 | |
XM_011523740.2 | XP_011522042.1 | 14 - 15 | 1201 | 53 | |
XM_011523741.2 | XP_011522043.1 | 14 - 15 | 1377 | 53 | |
XM_011523742.2 | XP_011522044.1 | 11 - 12 | 1083 | 53 | |
XM_011523743.2 | XP_011522045.1 | 12 - 13 | 1161 | 53 | |
XM_011523744.2 | XP_011522046.1 | 7 - 8 | 749 | 53 | |
XM_017024370.1 | XP_016879859.1 | 14 - 15 | 1266 | 53 | |
XM_017024371.1 | XP_016879860.1 | 13 - 14 | 1319 | 53 | |
GenBank mRNA | AB028962.1 | - | 3 - 4 | 342 | 53 |
AJ628446.1 | - | 14 - 15 | 1288 | 53 | |
AJ628447.1 | - | 13 - 14 | 1243 | 53 | |
AK124640.1 | - | 14 - 15 | 1377 | 53 | |
BC140870.1 | - | 14 - 15 | 1244 | 53 | |
BC140871.1 | - | 14 - 15 | 1244 | 53 | |
CR627427.1 | - | 13 - 14 | 1212 | 53 |