Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNB2
Entrez Gene ID
1948
Gene Name
ephrin B2
Gene Aliases
EPLG5, HTKL, Htk-L, LERK5
Chromosome Location
Chr.13: 106489731 - 106535040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004093.3 NP_004084.1 2 - 3 484 140
XM_017020406.1 XP_016875895.1 2 - 3 8745 140
GenBank mRNA BC069342.1 - 2 - 3 411 140
BC074856.2 - 2 - 3 411 140
BC074857.2 - 2 - 3 411 140
BC105955.1 - 2 - 3 429 140
BC105956.1 - 2 - 3 429 140
BC105957.1 - 2 - 3 429 140
L38734.1 - 2 - 3 439 140
U16797.1 - 2 - 3 415 140
U81262.1 - 2 - 3 415 140

View Full Product Details