The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018423.2 | NP_060893.2 | 4 - 5 | 710 | 77 |
XM_005253417.2 | XP_005253474.1 | 3 - 4 | 610 | 77 | |
XM_011520736.1 | XP_011519038.1 | 5 - 6 | 513 | 77 | |
XM_011520737.2 | XP_011519039.1 | 4 - 5 | 550 | 77 | |
XM_011520738.2 | XP_011519040.1 | 4 - 5 | 705 | 77 | |
GenBank mRNA | AF251059.1 | - | 4 - 5 | 710 | 77 |
AK313906.1 | - | 4 - 5 | 445 | 77 | |
AL353940.1 | - | 4 - 5 | 458 | 77 | |
AY563054.1 | - | 2 - 3 | 189 | 77 | |
BC093822.1 | - | 2 - 3 | 211 | 77 | |
BC093824.1 | - | 2 - 3 | 211 | 77 | |
KP729000.1 | - | 2 - 3 | 189 | 77 |