Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STYK1
Entrez Gene ID
55359
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
NOK, SuRTK106
Chromosome Location
Chr.12: 10618923 - 10674318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018423.2 NP_060893.2 7 - 8 1237 64
XM_005253417.2 XP_005253474.1 6 - 7 1137 64
XM_011520736.1 XP_011519038.1 8 - 9 1040 64
XM_011520737.2 XP_011519039.1 7 - 8 1077 64
XM_011520738.2 XP_011519040.1 7 - 8 1232 64
XM_017019579.1 XP_016875068.1 7 - 8 1020 64
GenBank mRNA AF251059.1 - 7 - 8 1237 64
AK313906.1 - 7 - 8 972 64
AL353940.1 - 7 - 8 985 64
AY563054.1 - 5 - 6 716 64
BC093822.1 - 5 - 6 738 64
BC093824.1 - 5 - 6 738 64
KP729000.1 - 5 - 6 716 64

View Full Product Details