The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079910.1 | NP_001073379.1 | 20 - 21 | 4528 | 106 |
XM_011538817.2 | XP_011537119.1 | 20 - 21 | 4471 | 106 | |
XM_011538818.2 | XP_011537120.1 | 20 - 21 | 4471 | 106 | |
XM_011538819.2 | XP_011537121.1 | 20 - 21 | 4471 | 106 | |
XM_011538823.2 | XP_011537125.1 | 20 - 21 | 4471 | 106 | |
XM_017020019.1 | XP_016875508.1 | 20 - 21 | 4471 | 106 | |
GenBank mRNA | CA502842.1 | - | 4 - 5 | 455 | 106 |