The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286732.1 | NP_001273661.1 | 5 - 6 | 661 | 71 |
NM_024719.3 | NP_078995.2 | 5 - 6 | 661 | 71 | |
XM_005268333.3 | XP_005268390.1 | 4 - 5 | 498 | 71 | |
XM_017020755.1 | XP_016876244.1 | 5 - 6 | 619 | 71 | |
XM_017020756.1 | XP_016876245.1 | 6 - 7 | 799 | 71 | |
GenBank mRNA | AB449887.1 | - | 5 - 6 | 564 | 71 |
AK026127.1 | - | 3 - 4 | 304 | 71 | |
AK300897.1 | - | 5 - 6 | 611 | 71 | |
BC033071.2 | - | 3 - 4 | 385 | 71 | |
BC062461.1 | - | 4 - 5 | 509 | 71 | |
BC080640.1 | - | 5 - 6 | 618 | 71 | |
BC112123.1 | - | 3 - 4 | 338 | 71 | |
BC112125.1 | - | 3 - 4 | 350 | 71 | |
BC143682.1 | - | 3 - 4 | 338 | 71 | |
CX788865.1 | - | 4 - 5 | 502 | 71 |