Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCF2L
Entrez Gene ID
23263
Gene Name
MCF.2 cell line derived transforming sequence like
Gene Aliases
ARHGEF14, DBS, OST
Chromosome Location
Chr.13: 112894378 - 113099739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112732.2 NP_001106203.2 13 - 14 1817 69
NM_001320815.1 NP_001307744.1 13 - 14 1849 69
NM_001320816.1 NP_001307745.1 13 - 14 2026 69
NM_001320817.1 NP_001307746.1 13 - 14 1849 69
NM_024979.4 NP_079255.4 13 - 14 1849 69
XM_011537482.1 XP_011535784.1 14 - 15 1806 69
XM_011537483.2 XP_011535785.1 14 - 15 1926 69
XM_011537484.1 XP_011535786.1 13 - 14 2253 69
XM_011537485.1 XP_011535787.1 13 - 14 1925 69
XM_011537486.2 XP_011535788.2 13 - 14 1941 69
XM_011537487.1 XP_011535789.1 13 - 14 1846 69
XM_011537488.1 XP_011535790.1 13 - 14 1685 69
XM_011537490.1 XP_011535792.1 13 - 14 2219 69
XM_011537491.1 XP_011535793.1 11 - 12 1634 69
XM_017020493.1 XP_016875982.1 13 - 14 1941 69
XM_017020494.1 XP_016875983.1 13 - 14 1941 69
XM_017020495.1 XP_016875984.1 13 - 14 1941 69
XM_017020496.1 XP_016875985.1 11 - 12 1634 69
XM_017020497.1 XP_016875986.1 13 - 14 2219 69
XM_017020498.1 XP_016875987.1 11 - 12 1634 69
XM_017020499.1 XP_016875988.1 13 - 14 1941 69
GenBank mRNA AB002360.1 - 13 - 14 1794 69
AB116074.1 - 13 - 14 1705 69
AB116075.1 - 8 - 9 1181 69
AK295047.1 - 13 - 14 69
AK295670.1 - 13 - 14 1659 69
AK315969.1 - 13 - 14 1817 69
BC011853.2 - 13 - 14 1809 69
BC020208.1 - 13 - 14 1809 69

View Full Product Details