Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MCF2L
- Entrez Gene ID
-
23263
- Gene Name
- MCF.2 cell line derived transforming sequence like
- Gene Aliases
-
ARHGEF14, DBS, OST
- Chromosome Location
-
Chr.13: 112894378 - 113099739 on Build GRCh38
View Full Product Details