Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
Chromosome Location
Chr.16: 85169512 - 85676206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134473.2 NP_001127945.1 3 - 4 481 90
NM_001278184.1 NP_001265113.1 3 - 4 535 90
NM_014615.3 NP_055430.1 4 - 5 754 90
XM_005255859.4 XP_005255916.3 5 - 6 2896 90
XM_005255860.3 XP_005255917.3 5 - 6 2903 90
XM_005255861.4 XP_005255918.3 4 - 5 2677 90
XM_005255863.4 XP_005255920.1 4 - 5 646 90
XM_005255864.4 XP_005255921.1 4 - 5 1378 90
XM_005255865.4 XP_005255922.1 3 - 4 1159 90
XM_011522965.2 XP_011521267.1 4 - 5 1145 90
XM_011522966.2 XP_011521268.1 4 - 5 612 90
XM_017023083.1 XP_016878572.1 4 - 5 904 90
GenBank mRNA BC037556.1 - 3 - 4 504 90
D80004.2 - 3 - 4 419 90

View Full Product Details