The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134473.2 | NP_001127945.1 | 4 - 5 | 684 | 86 |
NM_001278184.1 | NP_001265113.1 | 4 - 5 | 738 | 86 | |
NM_014615.3 | NP_055430.1 | 5 - 6 | 957 | 86 | |
XM_005255859.4 | XP_005255916.3 | 6 - 7 | 3099 | 86 | |
XM_005255860.3 | XP_005255917.3 | 6 - 7 | 3106 | 86 | |
XM_005255861.4 | XP_005255918.3 | 5 - 6 | 2880 | 86 | |
XM_005255863.4 | XP_005255920.1 | 5 - 6 | 849 | 86 | |
XM_005255864.4 | XP_005255921.1 | 5 - 6 | 1581 | 86 | |
XM_005255865.4 | XP_005255922.1 | 4 - 5 | 1362 | 86 | |
XM_011522965.2 | XP_011521267.1 | 5 - 6 | 1348 | 86 | |
XM_011522966.2 | XP_011521268.1 | 5 - 6 | 815 | 86 | |
XM_017023083.1 | XP_016878572.1 | 5 - 6 | 1107 | 86 | |
GenBank mRNA | BC037556.1 | - | 4 - 5 | 707 | 86 |
D80004.2 | - | 4 - 5 | 622 | 86 |