The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000264.3 | NP_000255.2 | 5 - 6 | 936 | 115 |
NM_001083602.1 | NP_001077071.1 | 5 - 6 | 1044 | 115 | |
NM_001083603.1 | NP_001077072.1 | 5 - 6 | 890 | 115 | |
NM_001083604.1 | NP_001077073.1 | 5 - 6 | 654 | 115 | |
NM_001083605.1 | NP_001077074.1 | 5 - 6 | 637 | 115 | |
NM_001083606.1 | NP_001077075.1 | 5 - 6 | 736 | 115 | |
NM_001083607.1 | NP_001077076.1 | 5 - 6 | 810 | 115 | |
GenBank mRNA | AB189436.1 | - | 5 - 6 | 1044 | 115 |
AB189437.1 | - | 5 - 6 | 890 | 115 | |
AB189438.1 | - | 5 - 6 | 656 | 115 | |
AB189439.1 | - | 5 - 6 | 639 | 115 | |
AB189440.1 | - | 5 - 6 | 809 | 115 | |
AB233424.1 | - | 4 - 5 | 586 | 115 | |
U43148.1 | - | 5 - 6 | 736 | 115 | |
U59464.1 | - | 5 - 6 | 748 | 115 |