The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000264.3 | NP_000255.2 | 6 - 7 | 1131 | 83 |
NM_001083602.1 | NP_001077071.1 | 6 - 7 | 1239 | 83 | |
NM_001083603.1 | NP_001077072.1 | 6 - 7 | 1085 | 83 | |
NM_001083604.1 | NP_001077073.1 | 6 - 7 | 849 | 83 | |
NM_001083605.1 | NP_001077074.1 | 6 - 7 | 832 | 83 | |
NM_001083606.1 | NP_001077075.1 | 6 - 7 | 931 | 83 | |
NM_001083607.1 | NP_001077076.1 | 6 - 7 | 1005 | 83 | |
GenBank mRNA | AB209495.1 | - | 3 - 4 | 580 | 83 |
AB233422.1 | - | 1 - 2 | 83 | 83 | |
U43148.1 | - | 6 - 7 | 931 | 83 | |
U59464.1 | - | 6 - 7 | 943 | 83 |