The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243254.1 | NP_001230183.1 | 1 - 2 | 366 | 109 |
NM_015213.3 | NP_056028.2 | 1 - 2 | 366 | 109 | |
XM_005252832.1 | XP_005252889.1 | 1 - 2 | 375 | 109 | |
XM_017017419.1 | XP_016872908.1 | 1 - 2 | 375 | 109 | |
GenBank mRNA | AB029014.1 | - | 1 - 2 | 326 | 109 |
AK295881.1 | - | 1 - 2 | 127 | 109 | |
AL117448.1 | - | 1 - 2 | 122 | 109 | |
BC071596.1 | - | 2 - 3 | 171 | 109 | |
BX401101.2 | - | 1 - 2 | 396 | 109 | |
DN999250.1 | - | 1 - 2 | 373 | 109 |