The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243254.1 | NP_001230183.1 | 7 - 8 | 1922 | 71 |
NM_015213.3 | NP_056028.2 | 7 - 8 | 1922 | 71 | |
XM_005252832.1 | XP_005252889.1 | 7 - 8 | 1931 | 71 | |
XM_011519953.2 | XP_011518255.1 | 1 - 2 | 54 | 71 | |
XM_017017419.1 | XP_016872908.1 | 7 - 8 | 1931 | 71 | |
GenBank mRNA | AB029014.1 | - | 7 - 8 | 1882 | 71 |
AK025499.1 | - | 2 - 3 | 533 | 71 | |
AK294016.1 | - | 6 - 7 | 1651 | 71 | |
AK295881.1 | - | 7 - 8 | 1683 | 71 | |
AK302478.1 | - | 4 - 5 | 1383 | 71 | |
AL117448.1 | - | 7 - 8 | 1678 | 71 | |
BC009354.2 | - | 3 - 4 | 650 | 71 | |
BC071596.1 | - | 8 - 9 | 1727 | 71 |