Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
Chromosome Location
Chr.11: 34150987 - 34358008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145804.2 NP_665803.2 4 - 5 1826 65
GenBank mRNA AK091560.1 - 1 - 2 44 65
AK095632.1 - 4 - 5 1073 65
AK291744.1 - 4 - 5 1827 65
BC093968.1 - 4 - 5 938 65

View Full Product Details