Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
Chromosome Location
Chr.11: 34150987 - 34358008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145804.2 NP_665803.2 9 - 10 2415 78
GenBank mRNA AK091560.1 - 6 - 7 633 78
AK095632.1 - 9 - 10 1662 78
AK291744.1 - 9 - 10 2416 78
BC093968.1 - 9 - 10 1527 78
BX640841.1 - 1 - 2 116 78

View Full Product Details