The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006289.3 | NP_006280.3 | 14 - 15 | 1930 | 98 |
GenBank mRNA | AB028950.2 | - | 14 - 15 | 1905 | 98 |
AF078828.1 | - | 14 - 15 | 1702 | 98 | |
AF177198.1 | - | 14 - 15 | 1666 | 98 | |
BC042923.1 | - | 14 - 15 | 1716 | 98 |