The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006289.3 | NP_006280.3 | 22 - 23 | 3229 | 99 |
GenBank mRNA | AB028950.2 | - | 22 - 23 | 3204 | 99 |
AF078828.1 | - | 22 - 23 | 3001 | 99 | |
AF177198.1 | - | 22 - 23 | 2965 | 99 | |
BC042923.1 | - | 22 - 23 | 3015 | 99 |