Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLN1
Entrez Gene ID
7094
Gene Name
talin 1
Gene Aliases
ILWEQ, TLN
Chromosome Location
Chr.9: 35697337 - 35732395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006289.3 NP_006280.3 56 - 57 7856 78
GenBank mRNA AB028950.2 - 56 - 57 7831 78
AF078828.1 - 56 - 57 7628 78
AF113217.1 - 1 - 2 78 78
AF177198.1 - 56 - 57 7592 78
BC042923.1 - 56 - 57 7642 78

View Full Product Details