The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052924.2 | NP_443156.2 | 10 - 11 | 1349 | 119 |
XM_005250772.3 | XP_005250829.1 | 11 - 12 | 1535 | 119 | |
XM_005250773.3 | XP_005250830.1 | 11 - 12 | 1532 | 119 | |
XM_006716502.3 | XP_006716565.1 | 11 - 12 | 1550 | 119 | |
XM_011516817.2 | XP_011515119.1 | 11 - 12 | 1553 | 119 | |
XM_011516818.1 | XP_011515120.1 | 11 - 12 | 2210 | 119 | |
XM_011516819.2 | XP_011515121.1 | 10 - 11 | 1394 | 119 | |
XM_011516820.2 | XP_011515122.1 | 10 - 11 | 1737 | 119 | |
XM_011516821.2 | XP_011515123.1 | 10 - 11 | 1741 | 119 | |
XM_011516822.2 | XP_011515124.1 | 10 - 11 | 1886 | 119 | |
XM_011516823.2 | XP_011515125.1 | 11 - 12 | 1554 | 119 | |
XM_017013013.1 | XP_016868502.1 | 10 - 11 | 1425 | 119 | |
XM_017013014.1 | XP_016868503.1 | 11 - 12 | 1551 | 119 | |
XM_017013015.1 | XP_016868504.1 | 10 - 11 | 1867 | 119 | |
XM_017013016.1 | XP_016868505.1 | 11 - 12 | 1535 | 119 | |
GenBank mRNA | AB067516.1 | - | 9 - 10 | 1283 | 119 |
AY082588.1 | - | 9 - 10 | 1291 | 119 | |
BC025767.1 | - | 10 - 11 | 1317 | 119 |