The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011538442.2 | XP_011536744.2 | 7 - 8 | 1446 | 59 |
XM_017019416.1 | XP_016874905.1 | 4 - 5 | 1039 | 59 | |
XM_017019418.1 | XP_016874907.1 | 4 - 5 | 467 | 59 | |
XM_017019419.1 | XP_016874908.1 | 2 - 3 | 271 | 59 | |
GenBank mRNA | AF116545.1 | - | 8 - 9 | 742 | 59 |