Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF4
Entrez Gene ID
6925
Gene Name
transcription factor 4
Gene Aliases
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location
Chr.18: 55222331 - 55635993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083962.1 NP_001077431.1 11 - 12 1539 78
NM_001243226.2 NP_001230155.2 12 - 13 1329 78
NM_001243227.1 NP_001230156.1 10 - 11 1373 78
NM_001243228.1 NP_001230157.1 11 - 12 1483 78
NM_001243230.1 NP_001230159.1 10 - 11 1015 78
NM_001243231.1 NP_001230160.1 9 - 10 973 78
NM_001243232.1 NP_001230161.1 7 - 8 995 78
NM_001243233.1 NP_001230162.1 7 - 8 936 78
NM_001243234.1 NP_001230163.1 4 - 5 568 78
NM_001243235.1 NP_001230164.1 4 - 5 568 78
NM_001243236.1 NP_001230165.1 4 - 5 584 78
NM_001306207.1 NP_001293136.1 10 - 11 1164 78
NM_001306208.1 NP_001293137.1 7 - 8 995 78
NM_003199.2 NP_003190.1 11 - 12 1539 78
XM_005266739.3 XP_005266796.2 10 - 11 1179 78
XM_005266741.3 XP_005266798.1 11 - 12 1139 78
XM_005266743.4 XP_005266800.1 10 - 11 1000 78
XM_005266744.3 XP_005266801.1 10 - 11 1120 78
XM_005266745.3 XP_005266802.1 10 - 11 982 78
XM_005266747.3 XP_005266804.1 9 - 10 976 78
XM_005266749.3 XP_005266806.1 7 - 8 767 78
XM_005266752.4 XP_005266809.1 7 - 8 1337 78
XM_005266754.3 XP_005266811.1 6 - 7 761 78
XM_005266755.4 XP_005266812.1 7 - 8 796 78
XM_005266761.3 XP_005266818.1 4 - 5 1080 78
XM_006722536.2 XP_006722599.1 11 - 12 1462 78
XM_006722537.2 XP_006722600.1 11 - 12 1095 78
XM_006722538.2 XP_006722601.1 10 - 11 1117 78
XM_011526158.2 XP_011524460.1 10 - 11 2093 78
XM_011526160.2 XP_011524462.1 6 - 7 1146 78
XM_017025934.1 XP_016881423.1 10 - 11 1176 78
XM_017025935.1 XP_016881424.1 10 - 11 1179 78
XM_017025936.1 XP_016881425.1 10 - 11 1179 78
XM_017025937.1 XP_016881426.1 10 - 11 933 78
XM_017025938.1 XP_016881427.1 11 - 12 1092 78
XM_017025939.1 XP_016881428.1 11 - 12 1142 78
XM_017025940.1 XP_016881429.1 11 - 12 1462 78
XM_017025941.1 XP_016881430.1 11 - 12 1095 78
XM_017025942.1 XP_016881431.1 11 - 12 1139 78
XM_017025943.1 XP_016881432.1 11 - 12 1142 78
XM_017025944.1 XP_016881433.1 10 - 11 1397 78
XM_017025945.1 XP_016881434.1 10 - 11 1400 78
XM_017025946.1 XP_016881435.1 10 - 11 1414 78
XM_017025947.1 XP_016881436.1 10 - 11 1000 78
XM_017025948.1 XP_016881437.1 10 - 11 1397 78
XM_017025949.1 XP_016881438.1 10 - 11 1117 78
XM_017025950.1 XP_016881439.1 10 - 11 1397 78
XM_017025951.1 XP_016881440.1 9 - 10 976 78
XM_017025952.1 XP_016881441.1 7 - 8 990 78
XM_017025953.1 XP_016881442.1 7 - 8 767 78
XM_017025954.1 XP_016881443.1 7 - 8 1351 78
XM_017025955.1 XP_016881444.1 6 - 7 1085 78
XM_017025956.1 XP_016881445.1 6 - 7 761 78
XM_017025957.1 XP_016881446.1 4 - 5 1080 78
GenBank mRNA AB209741.1 - 6 - 7 647 78
AK095041.1 - 7 - 8 995 78
AK096862.1 - 9 - 10 973 78
AK122765.1 - 11 - 12 1539 78
AK299169.1 - 10 - 11 1373 78
AK300038.1 - 4 - 5 514 78
AK300612.1 - 7 - 8 936 78
AK300636.1 - 6 - 7 666 78
AK301144.1 - 10 - 11 1150 78
AK315074.1 - 11 - 12 1471 78
AK316165.1 - 6 - 7 710 78
BC031056.1 - 12 - 13 1290 78
BC125084.1 - 10 - 11 953 78
BC125085.1 - 10 - 11 953 78
FR748210.1 - 10 - 11 927 78
FR748211.1 - 10 - 11 927 78
FR748212.1 - 8 - 9 747 78
FR748213.1 - 8 - 9 747 78
FR748214.1 - 9 - 10 855 78
FR748215.1 - 7 - 8 675 78
FR748216.1 - 10 - 11 921 78
FR748217.1 - 9 - 10 801 78
FR748218.1 - 5 - 6 537 78
FR748219.1 - 7 - 8 714 78
FR748220.1 - 4 - 5 447 78
FR748221.1 - 4 - 5 447 78
FR748222.1 - 4 - 5 447 78
FR748223.1 - 2 - 3 279 78
M74718.1 - 4 - 5 399 78
M74719.1 - 11 - 12 1126 78
X52079.1 - 8 - 9 792 78

View Full Product Details