Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
Chromosome Location
Chr.14: 21287977 - 21351316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020366.3 NP_065099.3 22 - 23 3615 71
XM_005267879.2 XP_005267936.1 13 - 14 2610 71
XM_005267880.2 XP_005267937.1 13 - 14 2577 71
XM_005267881.3 XP_005267938.1 11 - 12 2105 71
XM_011536978.1 XP_011535280.1 13 - 14 2607 71
XM_011536979.1 XP_011535281.1 14 - 15 2394 71
XM_011536980.1 XP_011535282.1 13 - 14 2265 71
XM_011536981.1 XP_011535283.1 11 - 12 2115 71
XM_011536982.1 XP_011535284.1 11 - 12 1770 71
XM_017021473.1 XP_016876962.1 11 - 12 2112 71
GenBank mRNA AF227257.1 - 11 - 12 2043 71
AF260257.1 - 13 - 14 2567 71
AF265666.1 - 11 - 12 2042 71
AF265667.1 - 11 - 12 1808 71
AJ417067.1 - 22 - 23 3615 71
AK301780.1 - 10 - 11 1673 71
BC039089.1 - 10 - 11 1658 71
BX571740.1 - 9 - 10 2547 71

View Full Product Details