The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020366.3 | NP_065099.3 | 9 - 10 | 1153 | 138 |
GenBank mRNA | AJ417067.1 | - | 9 - 10 | 1153 | 138 |