Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPH1
Entrez Gene ID
10808
Gene Name
heat shock protein family H (Hsp110) member 1
Gene Aliases
HSP105, HSP105A, HSP105B, NY-CO-25
Chromosome Location
Chr.13: 31134974 - 31163109 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286503.1 NP_001273432.1 8 - 9 1579 84
NM_001286504.1 NP_001273433.1 8 - 9 1440 84
NM_001286505.1 NP_001273434.1 6 - 7 1206 84
NM_006644.3 NP_006635.2 8 - 9 1579 84
XM_005266236.1 XP_005266293.1 8 - 9 2161 84
XM_011534887.2 XP_011533189.1 8 - 9 2161 84
XM_011534888.1 XP_011533190.1 6 - 7 1269 84
XM_017020361.1 XP_016875850.1 8 - 9 1503 84
XM_017020362.1 XP_016875851.1 8 - 9 2161 84
XM_017020363.1 XP_016875852.1 8 - 9 1503 84
XM_017020364.1 XP_016875853.1 6 - 7 1269 84
GenBank mRNA AB003333.1 - 8 - 9 1453 84
AB003334.1 - 8 - 9 1485 84
AF039695.1 - 8 - 9 1182 84
AK293955.1 - 8 - 9 1455 84
AK302294.1 - 6 - 7 857 84
AK302430.1 - 8 - 9 1440 84
AK302832.1 - 10 - 11 1530 84
AK314576.1 - 8 - 9 1503 84
BC037553.1 - 8 - 9 1481 84
BX648125.1 - 8 - 9 1417 84
BX648623.1 - 2 - 3 1787 84
D86956.1 - 8 - 9 1487 84

View Full Product Details