The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009932.2 | NP_001009932.1 | 4 - 5 | 773 | 67 |
NM_001009933.2 | NP_001009933.1 | 3 - 4 | 515 | 67 | |
NM_001009934.2 | NP_001009934.1 | 2 - 3 | 430 | 67 | |
NM_001303620.1 | NP_001290549.1 | 2 - 3 | 396 | 67 | |
NM_006730.3 | NP_006721.1 | 2 - 3 | 950 | 67 | |
XM_005277829.4 | XP_005277886.1 | 4 - 5 | 907 | 67 | |
XM_011531122.2 | XP_011529424.1 | 3 - 4 | 1456 | 67 | |
XM_017029332.1 | XP_016884821.1 | 4 - 5 | 1979 | 67 | |
GenBank mRNA | BC001561.1 | - | 2 - 3 | 314 | 67 |
BC028092.1 | - | 2 - 3 | 950 | 67 | |
BT019990.1 | - | 1 - 2 | 142 | 67 | |
L40823.1 | - | 3 - 4 | 475 | 67 | |
U06846.1 | - | 2 - 3 | 333 | 67 | |
X90392.1 | - | 3 - 4 | 936 | 67 |